missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min, Thermo Scientific Chemicals

Catalog No. p-7059390
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4450306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4450306 Supplier Thermo Scientific Chemicals Supplier No. 04450306

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13454-96-1 | Molecular Formula: Cl4Pt | Molecular Weight: 336.88 g/mol

Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370°C (decomposition)
Color Brown
Physical Form Crystalline Soluble
Quantity 5 g
UN Number UN2923
Solubility Information Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether.
Formula Weight 336.9
Percent Purity ≥99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 57% min
Odor Odorless
Sensitivity Hygroscopic
Density 4.303 g/mL
Assay (metals basis)
Chemical Name or Material Platinum (IV) chloride
Show More Show Less
Hazard Category H301-H314-H317-H334
Hazard Statement GHS H Statement
H301-H334-H314-H318-H317
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 236-645-1
RTECSNumber TP2275500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only