missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min

Catalog No. p-7076452
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4370303 1 g
AA4370306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4370303 Supplier Thermo Scientific Chemicals Supplier No. 04370303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process. | CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol

Platinum(IV) chloride is used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370°C (dec.)
Physical Form Crystalline
Quantity 1 g
UN Number UN2923
Solubility Information Soluble in water ( 587g/L ),acetone,slightly soluble in ethanol,ammonia. Insoluble in ether.
Formula Weight 336.9
Percent Purity ≥99.99%
Packaging Glass bottle
Sensitivity Hygroscopic
Specific Gravity 4.303
Density 4.303
Chemical Name or Material Platinum(IV) chloride
Show More Show Less
Hazard Category H301-H314-H317-H334
Precautionary Statement P260-P264b-P270-P272-P280-P285-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 236-645-1
RTECSNumber TP2275500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only