missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 1.0mm (0.04in) dia

Catalog No. AA42678BQ
Change view
Click to view available options
Quantity:
5 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42678BQ 5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42678BQ Supplier Thermo Scientific Chemicals Supplier No. 042678BQ

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Platinum Iridium wire
Name Note 1.0mm (0.04 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 5 cm
Odor Odorless
Weight ≈0.85g/5cm
Form Wire
MDL Number MFCD02091683

RUO – Research Use Only