missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium wire, 0.5mm (0.02in) dia, hard
Pt:Ir; 85:15 wt%
$261.26 - $261.26
Specifications
| Chemical Name or Material | Platinum Iridium wire |
|---|---|
| Molecular Formula | Pt:Ir; 85:15 wt% |
| MDL Number | MFCD02091683 |
| Packaging | Shrink-wrapped |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Platinum Iridium wire | |
| Pt:Ir; 85:15 wt% | |
| MFCD02091683 | |
| Shrink-wrapped |
RUO – Research Use Only