missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.5mm (0.02in) dia, hard

$248.90 - $905.60

Specifications

Chemical Name or Material Platinum Iridium wire
Name Note 0.5mm (0.02 in.) dia., Hard, Temper: as drawn
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 80:20 wt%
Odor Odorless
Weight ≈0.44g/10cm
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10054BS
View Documents
Thermo Scientific Chemicals
010054BS
10 cm
Each for $248.90
Only null left
 
AA10054BW
View Documents
Thermo Scientific Chemicals
010054BW
50 cm
Each for $905.60
Only null left
 
Description

Description

Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Iridium wire
Pt:Ir; 80:20 wt%
≈0.44g/10cm
MFCD02091683
0.5mm (0.02 in.) dia., Hard, Temper: as drawn
Odorless
Wire
Insoluble in water
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only