missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.2mm (0.008in) dia, annealed, 99.9+% (metals basis)

Catalog No. AA39526BS
Change view
Click to view available options
Quantity:
10 cm
50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39526BS 10 cm
AA39526BW 50 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39526BS Supplier Thermo Scientific Chemicals Supplier No. 039526BS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: Approx. 0.7g/10cm; Temper: soft; Formula: Pt:Ir; 80:20 wt% | CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum-iridium wire is used in platinum jewellery, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibres, in metrology and alloy is fabrication of metal microelectrodes. It is used in electrical stimulation of nervous tissue and electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Quantity 10 cm
Odor Odorless
Form Wire
Assay Percent Range >99.9%
Solubility Information Insoluble in water and acid.
Chemical Name or Material Platinum Iridium
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only