missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.25mm (0.01in) dia, annealed

Catalog No. AA43275BU
Change view
Click to view available options
Quantity:
25 cm
100 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43275BU 25 cm
AA43275BY 100 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43275BU Supplier Thermo Scientific Chemicals Supplier No. 043275BU

Form: Approx. 0.27g/25cm, Temper: soft | CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Quantity 25 cm
Odor Odorless
Form Wire
Chemical Name or Material Platinum Iridium
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only