missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.25mm (0.01in) dia, annealed, 99.9+% (metals basis)

Catalog No. AA39383BW
Change view
Click to view available options
Quantity:
10 cm
50 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39383BW 50 cm
AA39383BS 10 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA39383BW Supplier Thermo Scientific Chemicals Supplier No. 039383BW

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: approx. 0.11g/10cm,; Temper: soft | CAS: 7440-06-4 | Pt | 195.08 g/mol

Used in the fabrication of metal microelectrodes for electrical stimulation of nervous tissues and in electrophysiological recordings. Inoculating needles and leadwires for implants. Used in electrode materials, composing the gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver to Gray
Appearance Silver gray wire
Density 21.56g/cm3 at 20°C
Quantity 50 cm
Odor Odorless
Form Wire
Assay Percent Range >99.9%
Solubility Information Insoluble in water.
Chemical Name or Material Platinum Iridium
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only