missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)
Description
Platinum Iridium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used in the fabrication of micro electrodes for electrical stimulation of nervous tissue and electrophysiological recordings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Platinum Iridium foil |
| Quantity | 100 x 100 mm |
| Molecular Formula | Pt:Ir; 90:10 wt% |
| MDL Number | MFCD02091683 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.9% |
| Packaging | Tube |
RUO – Research Use Only