missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) iodide, Premion™, 99.99% (metals basis), Pt 43.0% min

Catalog No. p-7048946
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1217001 0.25 g
AA1217003 1 g
AA1217006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1217001 Supplier Thermo Scientific Chemicals Supplier No. 01217001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7790-39-8 | I2Pt | 448.89 g/mol

Platinum(II) iodide is used in pharmaceuticals and medical applications. It is also used to form cis and trans complexes with propan-1-amine. It is utilized in the catalytic hydration reaction of alkynes such as 2-hexyne, 4-methyl-2-pentyne, and 4,4dimethyl-2- pentyne . It is also used in the preparation of diiodo(2,9-dimethyl-1,10-phenanthroline)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7790-39-8
Molecular Formula I2Pt
Molecular Weight (g/mol) 448.89
MDL Number MFCD00011183
InChI Key ZXDJCKVQKCNWEI-UHFFFAOYSA-L
Synonym platinum ii iodide, platinum iodide pti2, platinum ii iodide, premion, platinum ii iodide,premion metalsbasis ,ptmin
IUPAC Name platinum(2+) diiodide
SMILES [I-].[I-].[Pt++]

Specifications

Melting Point 325°C (decomposition)
Physical Form Powder
Quantity 0.25 g
Odor Odorless
Solubility Information Insoluble in water,alkali iodides.
Formula Weight 448.9
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 43.0% min.
Assay (metals basis)
Chemical Name or Material Platinum(II) iodide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 232-204-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only