missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(II) Chloride 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 581°C |
| pH | 5.6 to 6.0 |
| Physical Form | Crystalline Powder |
| Quantity | 1 g |
| Formula Weight | 265.98 |
| Percent Purity | ≥95.0% (W) |
| Packaging | Glass Bottle |
| Chemical Name or Material | Platinum(II) Chloride |
Safety and Handling
| EINECSNumber | (1)-1145 |
| RTECSNumber | TP2275000 |
| TSCA | Yes |