missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) chloride, 99.9% (metals basis), Pt 73% min

Numéro de catalogue. AA1104403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA1104403 1 g
AA1104406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA1104403 Fournisseur Thermo Scientific Chemicals Code fournisseur 01104403

Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol

Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II). It is a well known chemotherapy drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 10025-65-7
Molecular Formula Cl2Pt
Molecular Weight (g/mol) 265.98
MDL Number MFCD00011181
InChI Key CLSUSRZJUQMOHH-UHFFFAOYSA-L
IUPAC Name platinum(2+) dichloride
SMILES [Cl-].[Cl-].[Pt++]

Spécifications

Melting Point 581°C (decomposition)
Color Brown-Green
Physical Form Crystalline
Quantity 1 g
Linear Formula PtCl2
UN Number UN3261
Merck Index 14,7528
Solubility Information Soluble in ammonia and hydrochloric acid; Insoluble in water,alcohol and ether.
Formula Weight 266
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pt 73% min.
Density 5.87 g/mL
Assay (metals basis)
Chemical Name or Material Platinum(II) chloride
Afficher plus de résultats Afficher moins de résultats
Hazard Category H314-H317-H334
Hazard Statement GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 233-034-1
RTECSNumber TP2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only