missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) acetylacetonate, 98%

Catalog No. p-4510688
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC369350010 1 g Glass bottle
AC369350050 5 g Glass bottle
AC369350250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC369350010 Supplier Thermo Scientific Chemicals Supplier No. 369350010
Only null left

CAS: 15170-57-7 | C10H14O4Pt | 393.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15170-57-7
Molecular Formula C10H14O4Pt
Molecular Weight (g/mol) 393.30
MDL Number MFCD00000028
InChI Key KLFRPGNCEJNEKU-FDGPNNRMSA-L
Synonym platinum ii acetylacetonate, pt acac 2, platinum bis acetylacetonate, acetylacetone platinum ii salt, platinum 2,4-pentanedionate, 2,4-pentanedione platinum ii derivative, bis acetylacetonato platinum, bis acetylacetonato platinum ii, 2,4-pentanedione, platinum ii
PubChem CID 10960186
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;platinum
SMILES [Pt++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 250.0°C to 252.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [CH3COCH=C(CH3)O]2Pt
Quantity 1 g
Sublimation Point 170 °C
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in ethanol, diethyl ether;, soluble in acetone, toluene, diluted acids
Formula Weight 393.31
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Platinum(II) acetylacetonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of damaging fertility or the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Cal
EINECSNumber 239-223-5

RUO – Research Use Only