Learn More
Platinum gauze, Unimesh N7433, expanded metal mesh, 0.34mm (0.013in) thick
CAS: 7440-06-4 | Pt | 195.08 g/mol
$771.44 - $2902.16
Chemical Identifiers
| CAS | 6-4-7440 |
|---|---|
| Molecular Formula | Pt |
| Molecular Weight (g/mol) | 195.08 |
| MDL Number | MFCD00011179 |
| InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
| Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
| PubChem CID | 23939 |
| ChEBI | CHEBI:33400 |
| IUPAC Name | platinum |
| SMILES | [Pt] |
Description
Platinum gauze is used in electronics industry for electrical contacts which might be subjected to high temperatures. It also acts as a catalyst in the production of nitric acid. It plays an essential role in the cracking processes in oil refineries. It can be coated on electrode surface by electroplating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6-4-7440 | |
| 195.08 | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| 23939 | |
| platinum |
| Pt | |
| MFCD00011179 | |
| black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
| CHEBI:33400 | |
| [Pt] |
Specifications
| 6-4-7440 | |
| 0.34mm Mesh | |
| ≈1.0g/25x25mm | |
| MFCD00011179 | |
| Soluble in hot aqua regia. | |
| [Pt] | |
| 195.08 | |
| CHEBI:33400 |
| 25 x 25 mm | |
| Odorless | |
| Pt | |
| black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| platinum | |
| 23939 | |
| Platinum gauze, Unimesh N7433, Expanded metal mesh, 0.34mm (0.013 in.) thick |
Safety and Handling
EINECSNumber : 231-116-1
RTECSNumber : TP2160000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only