missing translation for 'onlineSavingsMsg'
Learn More

Platinum foil, 0.127mm (0.005in) thick, Premion™, 99.99% (metals basis)

Form: Approx.1.7g/25 x 25mm | CAS: 7440-06-4 | Pt | 195.08 g/mol

$1209.23 - $3964.88

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42370FF
View Documents
Thermo Scientific Chemicals
042370FF
25 x 25 mm
Each for $1,209.23
Only null left
 
AA42370FI
View Documents
Thermo Scientific Chemicals
042370FI
50 x 50 mm
Each for $3,964.88
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6-4-7440
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]

Specifications

White
Silvery white foil
25 x 25 mm
99.99% (Metals basis)
Pt
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
[Pt]
195.08
CHEBI:33400
6-4-7440
21.45g/cm3 at 20°C
Odorless
Foil
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Platinum
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-116-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only