missing translation for 'onlineSavingsMsg'
Learn More

Platinum black

Catalog No. p-4824870
Change view
Click to view available options
Quantity:
0.1 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1207677 0.1 g
AA1207603 1 g
AA1207606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1207677 Supplier Thermo Scientific Chemicals Supplier No. 01207677

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Form: S.A. 10m2/g | CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum black is used in polymer electrolyte fuel cells, microbial fuel cells, and phosphoric acid fuel cell applications. It is also used as a recombination catalyst for hydrogen elimination. It is also used as a building block for sensor elements. It is used as a catalyst and as a gas absorbent. It is also useful for the electrochemical detection of hydrogen peroxide and nitrite in microchannel. It is useful for the anodic oxidation of ammonia in aqueous potassium hydroxide electrolyte.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Gray
Appearance Dark gray powder
Density 21.45g/cm3 at 20°C
Quantity 0.1 g
Odor Odorless
Form Powder
Solubility Information Soluble in aqua regia. Insoluble in water,mineral and organic acids.
Chemical Name or Material Platinum black
Hazard Category H228: Flammable solid.
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-116-1
RTECSNumber TP2160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only