missing translation for 'onlineSavingsMsg'
Learn More

Platinum Black, Powder, Spectrum™ Chemical

$770.40 - $7625.00

Chemical Identifiers

CAS 6-4-7440
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18-612-393
View Documents
Spectrum Chemical Mfg Cor
P1148250MG
250 mg Amber Glass Bottle
Each for $770.40
Only null left
 
18-612-392
View Documents
Spectrum Chemical Mfg Cor
P11481GM
1 g Amber Glass Bottle
Each for $1,902.97
Only null left
 
18-612-394
View Documents
Spectrum Chemical Mfg Cor
P11485GM
5 g Amber Glass Bottle
Each for $7,625.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6-4-7440

Specifications

6-4-7440
250 mg
Amber Glass Bottle
1
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.