missing translation for 'onlineSavingsMsg'
Learn More

Platinum on activated carbon, 5% Pt

Catalog No. p-42825
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC195230100 10 g Glass bottle
AC195230500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC195230100 Supplier Thermo Scientific Chemicals Supplier No. 195230100
Only null left

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Black
Quantity 10 g
Packaging Glass bottle
Particle Size Particle size distribution: <7 Micron: 10%, <28 Micron: 50%, <85 Micron: 90%
Form Powder
Fieser 01,982; 08,405; 04,393; 04,393; 04,393; 04,393
Formula Weight 195.08
Percent Purity 5% (Pt)
Chemical Name or Material Platinum on activated carbon, 5% Pt

RUO – Research Use Only