missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Platinum

Catalog No. 29809317G
Encompass
Change view
Click to view available options
Quantity:
1.7 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29809317G 1.7 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 29809317G Supplier Sigma Aldrich Supplier No. 2980931.7G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Platinum

Specifications

Melting Point 1772°C
Density 21.45 g/cm3 (literature)
Boiling Point 3827°C
Molecular Formula Pt
Linear Formula Pt
Quantity 1.7 g
MDL Number MFCD00011179
Molecular Weight (g/mol) 195.08
Percent Purity 99.9% trace metals basis