missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, 5% on carbon, sulfided, B106032-5, nominally 50% water wet
CAS: 7440-06-4 | Pt | 195.08 g/mol
$236.55 - $746.73
Chemical Identifiers
| CAS | 6-4-7440 |
|---|---|
| Molecular Formula | Pt |
| Molecular Weight (g/mol) | 195.08 |
| MDL Number | MFCD00011179 |
| InChI Key | BASFCYQUMIYNBI-UHFFFAOYSA-N |
| Synonym | black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion |
| PubChem CID | 23939 |
| ChEBI | CHEBI:33400 |
| IUPAC Name | platinum |
| SMILES | [Pt] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3654406
|
Thermo Scientific Chemicals
H3654406 |
5 g |
Each for $236.55
|
|
|||||
|
AAH3654414
|
Thermo Scientific Chemicals
H3654414 |
25 g |
Each for $746.73
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 6-4-7440 | |
| 195.08 | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| 23939 | |
| platinum |
| Pt | |
| MFCD00011179 | |
| black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
| CHEBI:33400 | |
| [Pt] |
Specifications
| Gray to Black | |
| Solid | |
| Pt | |
| black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion | |
| [Pt] | |
| 195.08 | |
| CHEBI:33400 | |
| Platinum, 5% on carbon, sulfided, B106032-5 |
| 6-4-7440 | |
| 5 g | |
| MFCD00011179 | |
| BASFCYQUMIYNBI-UHFFFAOYSA-N | |
| platinum | |
| 23939 | |
| Odorless |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
EINECSNumber : 231-116-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only