missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on carbon, dry

Catalog No. p-7081256
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1118603 1 g
AAA1118606 5 g
AAA1118614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1118603 Supplier Thermo Scientific Chemicals Supplier No. A1118603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts). Insoluble in water, mineral acids, and organic acids.

Notes
Do not store together with acids. Incompatible with Strong acids, strong oxidizing agents & organics Solvent vapors. Store under inert gas.

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 1 g
UN Number UN1325
Merck Index 14,7529
Solubility Information Soluble in boiling aqua regia (reacts),and molten alkali cyanides (reacts). Insoluble in water,mineral acids,and organic acids.
Chemical Name or Material Platinum, 5% on carbon, Dry
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 231-116-1
RTECSNumber TP2160000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only