missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on alumina powder, reduced

CAS: 7440-6-6

$857.95 - $857.95

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD03458043
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4422214
View Documents
Thermo Scientific Chemicals
04422214
25 g
Each for $857.95
Only null left
 
Description

Description

Platinum on Alumina (aluminum oxide) is catalyst material with applications in synthetic chemistry such as hydrogenation of functional groups. catalyst is recommended for selective hydrogenation reactions. The particle size of the catalyst is ideal for allowing fast separation from the reaction mixture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6-6-7440
MFCD03458043

Specifications

6-6-7440
25 g
MFCD03458043
Platinum, 5% on alumina powder
Powder
Powder
Insoluble in water,moderately soluble in strong mineral acids.
Videos
Safety and Handling

Safety and Handling

H228-H315-H319-H335 : Flammable solid. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only