missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 10% on carbon, dry

CAS: 7440-06-4 | Pt | 195.08 g/mol

Supplier:  Thermo Scientific Chemicals L0046906

 View more versions of this product

Catalog No. AAL0046906


Only null left
Explore available promotions

Description

Description

Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

Solubility
Insoluble in water.

Notes
Store in a cool location. Incompatible with oxidizing agents.
Specifications

Specifications

Platinum
6-4-7440
Odorless
MFCD00011179
14,7529
Insoluble in water.
[Pt]
195.08
CHEBI:33400
10% on carbon, dry
5 g
Pt
UN1325
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939

Chemical Identifiers

6-4-7440
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
CHEBI:33400
[Pt]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 231-116-1

RTECSNumber : TP2160000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only