missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 1% on gamma alumina powder, reduced

CAS: 7440-6-6

$519.00 - $519.00

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD00011179
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1179714
View Documents
Thermo Scientific Chemicals
01179714
25 g
Each for $519.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6-6-7440
MFCD00011179

Specifications

6-6-7440
25 g
Platinum, 1% on gamma alumina powder, Reduced
MFCD00011179
Insoluble in water.
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only