missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced

CAS: 7440-6-6

$192.42 - $2171.33

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD00011179
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA8910614
View Documents
Thermo Scientific Chemicals
08910614
25 g
Each for $192.42
Only null left
 
AA8910622
View Documents
Thermo Scientific Chemicals
08910622
100 g
Each for $487.22
Only null left
 
AA8910636
View Documents
Thermo Scientific Chemicals
08910636
500 g
Each for $2,171.33
Only null left
 
Description

Description

Platinum, 0.5% on 2.7-3.3mm (0.11-0.13in) alumina pellets is used for selective hydrogenations. It is also used in gas purifications, especially oxidation of carbon monoxide to carbon dioxide and hydrogen removal from oxygen or carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective hydrogenations; gas purifications: e
g
  • Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
  • Specifications

    Chemical Identifiers

    6-6-7440
    MFCD00011179

    Specifications

    6-6-7440
    25 g
    Platinum, 0.5% on alumina pellets, 2.7 to 3.3mm (0.11 to 0.13 in.), Reduced
    MFCD00011179
    Insoluble in water.
    Videos
    Safety and Handling

    Safety and Handling

    EINECSNumber : 215-691-6

    TSCA : Yes

    Recommended Storage : Ambient temperatures

    SDS
    missing translation for 'documents'

    missing translation for 'documents'

    RUO – Research Use Only