Learn More
Fisherbrand™ Platform Mats for Rockers and Wavers
For use with Fisherbrand™ Incubating Rockers, Open Air Rockers and Incubating Wavers.
$121.21
Specifications
Product Type Platform Mat
Material Rubber
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
02217735
View Documents Promotion Details
Fisherbrand™
80863652
Rockers
Each for $121.21
Only null left
Description
Description
  • Dimpled mat designed to hold tubes and vials of different sizes
  • Stacking tray (Cat. No. 02-217-736) attaches to Open Air Rocker unit to add second tier, mounts 3.5 in. (8.9cm) above lower tray, includes rubber mat and all hardware
Specifications
Specifications
Platform Mat
Rubber