missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Platform for Innova™ 2300/43/43R, Excella™ E10/E25/E25R

Catalog No. 05414369
Change view
Click to view available options
Holds:
12 x 2 L Erlenmeyer Flasks
15 x 1 L Erlenmeyer Flasks
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
05414369 12 x 2 L Erlenmeyer Flasks
05414368 15 x 1 L Erlenmeyer Flasks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05414369 Supplier Eppendorf™ Supplier No. AG2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Erlenmeyer flask dedicated platform for use with Innova™ 2300/43/43R, Excella™ E10/E25/E25R.

  • Platform for Innova™ 2300/43/43R, Excella™ E10/E25/E25R
  • 76 x 46 cm (30 x 18 in.)
  • Aluminum
  • Erlenmeyer flask dedicated platform

Specifications

Holds 12 x 2 L Erlenmeyer Flasks
Product Type Erlenmeyer Flask Platform
Material Aluminum
Dimensions (L x W) 76 x 46 cm (30 x 18 in.)
For Use With (Equipment) Innova™ 2300/43/43 R, Excella™ E10/E25/E25R