missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces

Catalog No. 12565841
Change view
Click to view available options
Color:
Clear
White
Product Type:
Microplate
Solid Module F8 (x12) in Frame
Surface Treatment:
Streptavidin
Total Volume (Metric) Well:
120 μL
400 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Surface Treatment Total Volume (Metric) Well
12565841 White Microplate Streptavidin 400 μL
1256715 Clear Microplate Streptavidin 400 μL
12565659 Clear Microplate Streptavidin 120 μL
1256716 Clear Solid Module F8 (x12) in Frame Streptavidin 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12565841 Supplier Thermo Scientific™ Supplier No. 436015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.

Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.

Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)

  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules

Specifications

Format Standard
Detection Method Chemiluminescence
Array 8 x 12
Packaging 15 Cases
Binding Type Affinity
Color White
Product Type Microplate
Material Plastic
Volume (Metric) Well 400 μL
No. of Wells 96
Volume (Metric) Working 100 μL
Surface Treatment Streptavidin
Total Volume (Metric) Well 400 μL
Well Shape Round, Flat Bottom (F-well)
No. per Case 15
No. per Pack 1
Plate Blocking Unspecified
Target Molecule Biotinylated
Show More Show Less