missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces

Catalog No. 12565659
Change view
Click to view available options
Color:
Clear
White
Product Type:
Microplate
Solid Module F8 (x12) in Frame
Surface Treatment:
Streptavidin
Total Volume (Metric) Well:
120 μL
400 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Surface Treatment Total Volume (Metric) Well
12565659 Clear Microplate Streptavidin 120 μL
1256715 Clear Microplate Streptavidin 400 μL
12565841 White Microplate Streptavidin 400 μL
1256716 Clear Solid Module F8 (x12) in Frame Streptavidin 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12565659 Supplier Thermo Scientific™ Supplier No. 436017

Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.

Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.

Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)

  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules

Specifications

Format Standard
Lid Without Lid
Detection Method Colorimetric
Packaging 15 Cases
Binding Type Affinity
Color Clear
Product Type Microplate
Material Polystyrene
Volume (Metric) Well 120 μL
No. of Wells 384
Volume (Metric) Working 50 μL
Sterility Non-sterile
Surface Treatment Streptavidin
Total Volume (Metric) Well 120 μL
Well Shape Square, Flat Bottom (F-well)
No. per Case 15
No. per Pack 1
Plate Blocking Unspecified
Quantity 15/Cs.
Target Molecule Biotinylated
Show More Show Less