Learn More
Gilson™ PLATEMASTERTM Lubrication Box
Prevent ill-fitting tips by regularly lubricating o-rings of laboratory pipettors.
Supplier:  Gilson™ F1077606
Catalog No. F1077606
Only null left
Explore available promotions
Description
Description
The Lubrication Box for O-rings is an accessory for the PLATEMASTERTM.
  • Lightly lubricate external o-ring between regular maintenance intervals to avoid ill-fitting tips
  • Ideal during periods of intensive use
Specifications
Specifications
Pipetter Accessory
ISO 9001
Polypropylene
PLATEMASTER™
1 Year
2.52 in
.22 lb
1
Not Autoclavable
Blue
Lubrication of O-rings
1
3.74 in
64 mm
.1 kg
Product Suggestions
Product Suggestions