missing translation for 'onlineSavingsMsg'
Learn More

BD Plated Media Trypticase™ Soy Agar

For isolation and cultivation of non-fastidious and fastidious microorganisms

$529.68 - $529.68

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Color Media Type Packaging Type Price Quantity  
Catalog Number Mfr. No. Quantity Color Media Type Packaging Type Price Quantity  
B4392385
View Documents
BD
292385
100 mL Light Beige Trypticase™ Soy Agar Bottle
Case of 10 Each for $529.68
Only null left
 
Description

Description

Specifications

Specifications

ISO/DIS, 11014
Liquid
Trypticase™ Soy Agar
2°C to 8°C
United States
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'