missing translation for 'onlineSavingsMsg'
Learn More

BD Plated Media Trypticase™ Soy Agar

Catalog No. B4392385
Change view
Click to view available options
Quantity:
100 mL
Color:
Light Beige
Media Type:
Trypticase™ Soy Agar
Packaging Type:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color Media Type Packaging Type
B4392385 100 mL Light Beige Trypticase™ Soy Agar Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4392385 Supplier BD Supplier No. 292385
Only null left

For isolation and cultivation of non-fastidious and fastidious microorganisms

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Content And Storage 2°C to 8°C
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar
Packaging Type Bottle
Quantity 100 mL