Learn More
Kinematica™ Plate Stand ST-P15/320 for Polytron™ 1300D Homogenizer
Description
With height-adjustable holder for drive support stainless steel for use with Polytron™ PT 1300 D drive.
Specifications
Specifications
| Description | Stainless steel stand with base, height-adjustable bracket for supporting the drive and base mounting |
| For Use With (Equipment) | PT 1300 D Disperser |
| Includes | The safety ring prevents the uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height. |
| Model | ST-P 15/320 |
| Product Type | Plate Stand |