missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Count Agar

Catalog No. p-4523948
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
20mm x 150mm Tube
85mm Monoplate
Bottle - 200mL
Bottle - 500mL
Media Type:
Plate Count Agar
Manufacturing Origin:
United States
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Media Type Manufacturing Origin
R01685 10/Pk. 85mm Monoplate Plate Count Agar United States
R09642 20/Pk. 20mm x 150mm Tube Plate Count Agar United States
R112590 10/Pk. Bottle - 200mL Plate Count Agar United States
R112591 10/Pk. Bottle - 500mL Plate Count Agar United States
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R01685 Supplier Thermo Scientific™ Supplier No. RE01685
Only null left

Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.

Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.

This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications

Format 85mm Monoplate
Manufacturing Origin United States
Media Type Plate Count Agar
Product Type Agar
Quantity 10/Pk.