missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasticoid Flanged Red Rubber Stoppers for Serum and Vaccine Bottles
Permit insertion of hypodermic needles
Supplier: Plasticoid 20190
Description
- Comprised of dry natural rubber
- Flange O.D.: 21mm
- Plug Top O.D.: 16mm
- Plug Bottom O.D.: 14mm
- Length: 19mm
- Fits 5 through 125mL size bottles
Specifications
| Natural | |
| 1.4 cm | |
| Rubber | |
| 1.9 cm |
| Flanged Stopper | |
| 1.62 cm | |
| Serum and vaccine bottles |