Learn More
United Scientific™ Plastic Wave Helix
Catalog No. S000875
Change view
Click to view available options
Length (English):
8 ft.
Catalog No. Length (English)
S000875 8 ft.
1 options
Prop 65
Catalog No. S000875 Supplier United Scientific™ Supplier No. UNUNPSLKYC
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Prop 65
This Plastic Wave Helix is a great demonstration to illustrate wave properties.
Holding the spring vertically demonstrates longitudinal waves when oscillated up and down. When two people hold each end and move one end up and down this visualizes a transverse wave. Made from PVC plastic. The overall dimensions are 3 1/4 in. in diameter and 2 1/2 in. tall.
  • Demonstrate transverse and longitudinal waves
  • Colorful design
  • Can safely stretch up to 8 ft.
Note: Colors may vary.
Specifications
Diameter (English) Coil 3 1/4 in.
Length (English) 8 ft.
Product Type Wave Demonstrator Spring Set
Height (English) 2 1/2 in.