missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Plastic Vernier Caliper

Catalog No. S12912
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S12912 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S12912 Supplier Eisco™ Supplier No. PH0078B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Columbus-type Plastic Vernier Caliper with dual scale.

  • Enable user to read inside, outside or depth measurements.
  • With thumb wheel for easy motion of jaws.
  • Metric scale graduated 0 to 12 cm x 0.1 mm.
  • Imperial scale graduated to 5 x 1/16 in.
  • Available in plastic case.

Specifications

Grade 6 to Undergraduate
Dimensions (L x W) 5 x 0.06 in.
Includes Thumb wheel for easy motion of jaws
Material Plastic
Scale Dual
Class Size 1 Lab Group
Product Type Vernier Caliper
Description Vernier caliper