missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Plastic Tweezers

Catalog No. S15680
Change view
Click to view available options
Length (Metric):
10.16 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S15680 10.16 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15680 Supplier United Scientific™ Supplier No. P10121
Edge

Economical tweezers for use in many applications.

  • These polypropylene tweezers are 4 in. long and feature serrated tips.
  • Non-sterile
  • Autoclavable
  • Sold as a pack of 12

Specifications

Product Type Tweezers
Length (English) 4 in.
Length (Metric) 10.16 cm
Material Polypropylene
Quantity 12/Pk.
Style Standard
Tip Style Serrated