missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Youngstown Barrel & Drum Plastic Tight Head Drums
High-density polyethylene containers for transport of hazardous solids or liquids
Supplier: Youngstown Barrel & Drum YBD1410
Description
- Made of chemical-resistant high-density polyethylene
- Tight-head drums for transport of hazardous liquids
Chemical Identifiers
| Plastic Tight Head Drums | |
| Plastic | |
| Tight |
| For Transport of Hazardous Solids or Liquids | |
| 50 |
Specifications
| UN 1H1/Y2.0/150 | |
| 15 gal. | |
| Natural | |
| Plastic | |
| Tight | |
| 20.6 in. | |
| 16 in. | |
| 20.94 lb. |
| Plastic Tight Head Drums | |
| 56.7 L | |
| For Transport of Hazardous Solids or Liquids | |
| 50 | |
| 2 Handles | |
| 52.32 cm | |
| 48.9 mm | |
| 9.5 Kg |