missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Plastic Test Tube Rack, Round, 12-Tube

Catalog No. S15810
Change view
Click to view available options
Positions:
28
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S15810 28
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15810 Supplier United Scientific™ Supplier No. 77704
Only null left

Economical way to safely store test tubes.

  • This circular 12 place polypropylene test tube stand base has matching wells and 16 drying pins surrounded by a trough to catch drippings.
  • Four 25 mm diameter holes and eight 19 mm diameter holes
  • Pre-assembled

Specifications

Autoclavable Autoclavable
Color White
Holds Test Tubes
Material Polypropylene
Positions 28
Shape Round
Stackable No
Sterility Non-sterile
For Use With (Equipment) Test Tubes
Includes One Rack
Product Type Test Tube Rack
Shelves None
Array 12 in a Circle
Show More Show Less