missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Plastic Test Tube Rack, 3-Tier

Catalog No. S15979
Change view
Click to view available options
Positions:
12
18
20
31
36
40
62
For Use With (Equipment):
13 mm Tubes
15 mm Tubes
16 mm Tubes
18 mm Tubes
20 mm Tubes
25 mm Tubes
32 mm Tubes
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions For Use With (Equipment)
S15979 18 13 mm Tubes
S15980 31 13 mm Tubes
S15981 18 25 mm Tubes
S16043 18 15 mm Tubes
S16081 62 16 mm Tubes
S15958 12 25 mm Tubes
S16006 36 25 mm Tubes
S15982 31 16 mm Tubes
S15729 40 20 mm Tubes
S15731 12 32 mm Tubes
S15991 62 13 mm Tubes
S15851 12 18 mm Tubes
S15799 20 20 mm Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. S15979 Supplier United Scientific™ Supplier No. 77714
Only null left

Can be disassembled to store flat

  • Polypropylene racks with a 3-tier design provide a clear view of the tube contents and convenient insertion and removal of tubes.
  • Available in multiple diameters and configurations.

Specifications

Autoclavable Autoclavable
Color White
Holds Test Tubes
Material Polypropylene
Positions 18
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 13 mm Tubes
Includes 4 Racks
Product Type Test Tube Rack (3-tier)
Shelves 3 Tiers
Array 3 x 6
Show More Show Less