missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Plastic Test Tube Rack, 12-Tube

Catalog No. S15788
Change view
Click to view available options
Positions:
12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
S15788 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15788 Supplier United Scientific™ Supplier No. TTHP03
Only null left

Popular design breaks down flat for easy storage.

  • Sturdy polypropylene rack will hold 12 test tubes up to 20 mm in diameter.
  • Easy to assemble, and chemical resistant.

Specifications

Autoclavable Autoclavable
Color White
Holds Test Tubes
Material Polypropylene
Positions 12
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) 20 mm Tubes
Includes One Rack
Product Type Test Tube Rack
Shelves 2 Tiers
Array 2 x 6
Show More Show Less