Learn More
Legend PLASTIC PULP DENSITY CONTAINER
Catalog No. NC9726404
Encompass
Plastic container for Marcy Pulp Density Scale. Includes handle and overflow slot. Volume: 1 Liter
Catalog No. NC9726404 Supplier Legend Supplier No. 18386
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Plastic container for Marcy Pulp Density Scale. Includes handle and overflow slot. Volume: 1 Liter