missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Essentials Plastic Pasteur Pipettes

Catalog No. 03391280
Change view
Click to view available options
Length (Metric):
146 mm
Material:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Material
03391280 146 mm Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03391280 Supplier Cole-Parmer™ Essentials Supplier No. 0439511

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A safe alternative to glass pipettes

  • Manufactured from medical-grade polystyrene in an ISO Class 8 cleanroom
  • Gamma irradiated, and RNase, DNase, and pyrogen free

Disposable polystyrene (PS) Pasteur pipettes offer the aspirating precision of glass, with the safety and benefits of plastic. Ideal for use with aspirating bulbs or vacuum applications to remove liquid waste from flasks, dishes, plates, and tubes. The long flexible stem can be bent to draw liquid from narrow or small volume tubes. Supplied individually wrapped.

Specifications

Material Plastic
Sterility Sterile
Graduated No
Length (English) 5.75 in.
Packaging Type Individually wrapped
Color Clear
Length (Metric) 146 mm
Type Pasteur Pipet
Disposable Single-use