missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plastic Pail with Tear Tab Lid

Catalog No. 19058153
Change view
Click to view available options
Capacity (English):
1 gal.
2 gal.
3.5 gal.
5 gal.
Handle Material:
10 Gauge Galvanized Wire
12 Gauge Galvanized Wire
9 Gauge Galvanized Wire
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Handle Material
19058153 5 gal. 9 Gauge Galvanized Wire
19058150 1 gal. 12 Gauge Galvanized Wire
19058151 2 gal. 10 Gauge Galvanized Wire
19058152 3.5 gal. 9 Gauge Galvanized Wire
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19058153 Supplier Fisherbrand™ Supplier No. 2300FB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Durable HDPE pail with reusable tear tab lid.

  • Prime High Density Polyethylene (HDPE) construction
  • Round, open head pail
  • Reusable tear tab lid
  • Galvanized wire handle
  • White

Specifications

Product Type New 5 gallon Plastic Pail tear tab lid
Capacity (Metric) 18.92 L
Material HDPE
Autoclavable Not Autoclavable
Capacity (English) 5 gal.
Certifications/Compliance Non-UN
Color White
Description New 5 gallon Plastic pail w/tear tab lid, Child safety label
Diameter (English) Wheel 11 15/16 in.
Dimensions 11-15/16 x 11-15/16 x 14-1/2 in.
For Use With (Application) Packaging and Shipping
Handle Material 9 Gauge Galvanized Wire
Height (English) 14-1/2 in.
Includes Tear tab Lid
Length (English) 11-15/16 in.
Quantity 1 Ea.
Width (English) 11-15/16 in.
Show More Show Less