Learn More
United Scientific™ Plastic Insulated Copper Wire, 100-Foot Roll
Useful in a wide variety of electricity demonstrations.
Supplier:  United Scientific™ WCP22W
 View more versions of this product
Catalog No. S15772
Only null left
Description
Description
  • 22-gauge (SWG) copper wire.
  • Supplied in a 100-foot roll.
  • Available with white, black, or red plastic insulation.
Specifications
Specifications
White
Wire
Copper
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
Warning: Cancer and reproductive harm