missing translation for 'onlineSavingsMsg'
Learn More

Eldon James Plastic Hose Barb Coupler

Eldon James features straight couplers and reduction couplers that are perfect for conjoining tubes of any kind. Safe for use in medical practices, our straight couplers and reduction couplers feature innovative single barb technology to stay firmly entrenched in the tubing.

$537.61 - $1588.07

Specifications

Description Class 7 cleanroom manufactured
Material Kynar PVDF
Color Natural
View More Specs

Products 11
Catalog Number Mfr. No. Product Type Hose Barb Price Quantity  
Catalog Number Mfr. No. Product Type Hose Barb Price Quantity  
01000824
View Documents
Eldon James
C108NK7QC
Reduction Coupler 10 Barb (5/8 in.) to 8 Barb (1/2 in.)
Pack of 100 Each for $1,019.57
Only null left
 
01000825
View Documents
Eldon James
C1210NK7QC
Reduction Coupler 12 Barb (3/4 in.) to 10 Barb (5/8 in.)
Pack of 100 Each for $1,285.29
Only null left
 
01000826
View Documents
Eldon James
C126NK7QC
Reduction Coupler 12 Barb (3/4 in.) to 6 Barb (3/8 in.)
Pack of 100 Each for $1,161.71
Only null left
 
01000827
View Documents
Eldon James
C128NK7QC
Reduction Coupler 12 Barb (3/4 in.) to 8 Barb (1/2 in.)
Pack of 100 Each for $1,223.50
Only null left
 
01000828
View Documents
Eldon James
C1610NK7QC
Reduction Coupler 16 Barb (1 in.) to 10 Barb (5/8 in.)
Pack of 100 Each for $1,588.07
Only null left
 
01000829
View Documents
Eldon James
C64NK7QC
Reduction Coupler 6 Barb (3/8 in.) to 4 Barb (1/4 in.)
Pack of 100 Each for $716.79
Only null left
 
01000830
View Documents
Eldon James
C84NK7QC
Reduction Coupler 8 Barb (1/2 in.) to 4 Barb (1/4 in.)
Pack of 100 Each for $883.64
Only null left
 
01000831
View Documents
Eldon James
C86NK7QC
Reduction Coupler 8 Barb (1/2 in.) to 6 Barb (3/8 in.)
Pack of 100 Each for $914.54
Only null left
 
01000832
View Documents
Eldon James
C012NK7QC
Straight Coupler 12 Barb (3/4 in.)
Pack of 100 Each for $1,347.11
Only null left
 
01000833
View Documents
Eldon James
C03NK7QC
Straight Coupler 3 Barb (3/16 in.)
Pack of 100 Each for $537.61
Only null left
 
01000834
View Documents
Eldon James
C06NK7QC
Straight Coupler 6 Barb (3/8 in.)
Pack of 100 Each for $803.32
Only null left
 
Description

Description

  • Our medical and pharmaceutical grade plastic tube couplers are RoHS (Restriction of Hazardous Substances) compliant, meet USP Class VI requirements, and are animal derivative-free.
  • Transportation and automotive barb couplings meet automotive standards and approvals.
  • Eldon James specializes in producing safe, efficient, and high quality products, all of which are made from Eco-friendly resins that will not leach pollutants or contaminate water, liquBarbs, gases, soil, or even other recyclable materials.
Specifications

Specifications

Class 7 cleanroom manufactured
Kynar PVDF
Natural
Videos
SDS
Documents

Documents