missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plastic Drum, 100L, for HyPerforma™ Single-Use Mixer DS 300, with Conical Insert

The 100 L Plastic Drum with Conical Insert is designed for use with the Thermo Scientific HyPerforma Single-Use Mixer (S.U.M.) DS 300 and is easily moved to cold rooms for storage.

Supplier:  Thermo Scientific™ SH3095902

Catalog No. SH3095902


Only null left
Explore available promotions

Description

Description

The 100 L Plastic Drum with Conical Insert adds speed and convenience to your workflow. The drum is designed for use with the Thermo Scientific™ HyPerforma™ Single-Use Mixer (S.U.M.) DS 300 and is easily moved to cold rooms for storage.

Key features:
• Standard plastic drums available in 50, 100, 200, and 300 L sizes
• Lightweight for ease of mobility
• Applicable to open- and closed-top mixing applications
• Economical
• Bottom drain for easy transfer
• 5:1 turn down ratio in conjunction with docking station and a bioprocess container bag or liner

Optional:
• Dollies for the drums are available in two sizes: one for 50 to 200 L drums and another for 300 L drums. These drums should be used with the corresponding optional drum positioner that is available with the docking station.

Required accessories:
• HyPerforma Single-Use Mixer DS 300 docking station
• HyPerforma Single-Use Mixer DS 300 bioprocess container bags for closed-top mixing or liners for open-top mixing

Recommended for:
• Powder-liquid or liquid-liquid mixing applications in single-use
• High-throughput mixing and storage

Specifications

Specifications

100 L
100 L
HyPerforma
23.5 x 29.75 in. (60 x 76 cm)
Bottom drain: 4 in. (10.2 cm)
DS 300 Single-Use Mixer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions