missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Plastic Disposable Generator Probe

12 mm Fisherbrand Plastic Disposable Generator Probe significantly reduces the complexity of disassembly and maintenance compared to traditional stainless-steel probes. Made from durable plastic, these tips are cost-effective for single use, minimizing the risk of cross-contamination.
$1,686.20 - $6,113.48
Specifications
| Product Type | Probe |
|---|---|
| Length (Metric) Probe | 12 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Closure Color | Blue |
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
15341134
![]() |
Fisherbrand™
OMNF301250 |
25/Pk. |
Pack for $1,686.20
|
|
|||||
15341135
![]() |
Fisherbrand™
F321250 |
50/Pk. |
Pack for $3,218.43
|
|
|||||
15341136
![]() |
Fisherbrand™
F341250 |
100/Pk. |
Pack for $6,113.48
|
|
|||||
Description
- Suitable for homogenization where sample size or sample hardness dictates that processing with the 7 mm tip is prohibitive.
- Significantly reduces cross-contamination and is virtually maintenance-free
- Suitable for PCR, RNA, and DNA extraction
- Compatible with Fisherbrand™ 150 Handheld Homogenizer, Fisherbrand™ 850 Homogenizer, Omni Tissue Homogenizer, Omni THQ, and Omni GLH 850 homogenizer
- Provides a cost-effective solution for researchers homogenizing samples where contaminants can affect downstream assays
Specifications
| Probe | |
| Autoclavable | |
| Blue |
| 12 mm | |
| Polypropylene |