Learn More
Research Products Corporation Coplin Staining Jars, Plastic for Microwave Applications, 6 per Case
Catalog No. 50212282
Encompass
Popular jars for staining and processing microscope slides. Fixed slides can be stained and dried without removing from jar. Plastic coplin jar and cap are molded from thick high density polyethylene that is microwave transparent. Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1 x 3 inch slides.
Catalog No. 50212282 Supplier Research Products Corporation Supplier No. RPI 144208
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Popular jars for staining and processing microscope slides. Fixed slides can be stained and dried without removing from jar. Plastic coplin jar and cap are molded from thick high density polyethylene that is microwave transparent. Polyethylene lid is vented for use in microwave applications. Internal grooves hold up to ten standard 1 x 3 inch slides.